Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Bottle Closures
Screw cap for round bottle series 321 COEX
Screw cap for round bottle series 321 COEX
KAUT805-86734
:
Kautex
Screw cap for round bottle series 321 COEX
KAUT805-86734
:
Kautex
Thread:
50 mm
Colour:
White
Description:
With PA/PE/PA insert and tamperevident ring
Material:
HDPE
Pk:
800
For bottles:
250 - 1000 ml