Assistent® Stirring Spatula
Stirring spatula with crossbar, polystyrene, -, Clear
Catalog # HECH40734010
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Designed for several uses, such as mixing samples and solutions in test tubes, resolution of substrate, additions of small reagent volumes, or releasing blood clots from the side of test tube walls.
- Stirring rod for a wide variety of laboratory applications
Made from polystyrene, crystal clear.
Specifications
- Sterile:-
- Colour:Clear
- Description:Stirring spatula with crossbar, polystyrene