ContactUsLinkComponent
Luer lock valve adaptors
Luer lock valve adaptors
Catalog # REST21283
Supplier:  Restek
undefined
Luer lock valve adaptors
Catalog # REST21283
Supplier:  Restek
Specifications
  • Description:
    Luer lock valve adaptor for syringes
  • Pk:
    2
Specifications
About this item
These Luer lock valve adaptors are suitable for connecting a Luer lock fitting to a Rheodyne or Valco valve.