XERT-500 column test mix (8 components)
Catalog # REST35247
Supplier: Restek
CAS Number:
Specifications
- Pk:1
- Description:XERT-500 column test mix, 500 µg/ml each in toluene
- Pk Info:1 ml/ampoule
Specifications
About this item
This contains (C9:0 FAME) methyl nonanoate, (C13) n-tridecane, (C15) n-pentadecane, acenaphthylene, 4-chlorophenol, n-decylamine, 1,6-hexanediol and 1-undecanol (112-42-5).