VOA calibration mix #3 (10 components)
REST30008
: Restek
:
- Concentration:2000 μg/ml each in P and T methanol
- Pk:1
- Description:VOA calibration mix #3
- Pk Info:1 ml/ampoule
Mix contains: Carbon tetrachloride (56-23-5), Chlorobenzene (108-90-7), Chloroform (67-66-3), 1,1-Dichloroethane (75-34-3), 1,1-Dichloroethene (75-35-4), 1,2-Dichloropropane (78-87-5), Methylene chloride (dichloromethane) (75-09-2), 1,1,2-Trichloroethane (79-00-5), Trichloroethene (79-01-6), m-Xylene (108-38-3).
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.