ContactUsLinkComponent
8095 calibration mix B (7 components)
8095 calibration mix B (7 components)
Catalog # REST31608
Supplier:  Restek
CAS Number:  
undefined
8095 calibration mix B (7 components)
Catalog # REST31608
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    8095 calibration mix B, 5000 μg/ml each in acetonitrile (3,5-dinitroaniline at 1000 μg/ml)
  • Pk Info:
    1 ml/ampoule
Specifications
About this item
The mix consists of, 3,5-dinitroaniline, nitrobenzene, nitroglycerin, 2-nitrotoluene, 3-nitrotoluene, 4-nitrotoluene, PETN.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.