8095 calibration mix A (10 components)
REST31607
: Restek
:
- Pk:1
- Description:8095 calibration mix A, 1000 μg/ml each in acetonitrile
- Pk Info:1 ml/ampoule
The mix consists of, 2-amino-4,6-dinitrotoluene, 4-amino-2,6-dinitrotoluene, 1,3-dinitrobenzene, 2,4-dinitrotoluene, 2,6-dinitrotoluene, HMX, RDX, tetryl, 1,3,5-trinitrobenzene, 2,4,6-trinitrotoluene.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.