Blood alcohol mix resolution control standard (8 components)
Catalog # REST36256
Supplier: Restek
CAS Number:
Specifications
- Concentration:0,100 g/dl each in water
- Pk:1
- Description:Blood alcohol mixture resolution control standard
- Pk Info:1 ml/ampoule
Specifications
About this item
The blood alcohol mix resolution is used to verify the retention time for each compound normally included in a blood alcohol test and to verify that the compounds are resolved from and do not interfere with one another.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.