Order Entry
Slovenia
ContactUsLinkComponent
MCPP, Restek
MCPP, Restek
Catalog # REST32271
Supplier:  Restek
CAS Number:  
undefined
MCPP, Restek
Catalog # REST32271
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    1000 μg/ml in methanol
  • Pk:
    1
  • CAS No.:
    93-65-2
  • Description:
    MCPP
  • Formula:
    C₁₀H₁₁ClO₃
  • Description:
    MCPP
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.