Order Entry
Slovenia
Orders LinkContactUsLinkComponent
PCB kit #1

PCB kit #1

PCB kit #1, 1000 µg/ml each in hexane, 1 ml/ampoule, 1
Catalog # REST32089
Supplier:   Restek
PCB kit #1
Catalog # REST32089
Supplier:  Restek
CAS Number:  
PCB kit #1
PCB kit #1

PCB kit #1, 1000 µg/ml each in hexane, 1 ml/ampoule, 1

CAS Number:  
Price
383,00 €
Log in for availability
1 * 1 items
Product Details & Documents

This kit contains Aroclor 1016, Aroclor 1221, Aroclor 1232, Aroclor 1242, Aroclor 1248, Aroclor 1254, Aroclor 1260.

  • Aroclor standards may exhibit lot-to-lot variation

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Concentration:
    1000 µg/ml each in hexane
  • Pk:
    1
  • Description:
    PCB kit #1
  • Pk Info:
    1 ml/ampoule

Specifications