Order Entry
Slovenia
Orders LinkContactUsLinkComponent
608 complete kit

608 complete kit

608 complete kit, 1 ml each of the mixtures, 1 mL
Catalog # REST32060
Supplier:   Restek
608 complete kit
Catalog # REST32060
Supplier:  Restek
CAS Number:  
608 complete kit
608 complete kit

608 complete kit, 1 ml each of the mixtures, 1 mL

CAS Number:  
Price
425,50 €
Log in for availability
1 * 1 mL
Product Details & Documents

This kit contains 1 ml of each of these mixtures 608 calibration mix, Aroclor 1016, Aroclor 1221, Aroclor 1232, Aroclor 1242, Aroclor 1248, Aroclor 1254, Aroclor 1260, toxaphene, chlordane.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    608 complete kit
  • Pk Info:
    1 ml each of the mixtures

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.