Herbicide reference standard, Dicamba
Catalog # REST32247
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml in methanol
- Pk:1
- CAS No.:1918-00-9
- Description:Dicamba
- Formula:C₈H₆Cl₂O₃
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.