Herbicide reference standard, 2,4-DB [4-(2,4-Dichlorophenoxy)butyric acid]
Catalog # REST32241
Supplier: Restek
CAS Number:
Specifications
- Concentration:1,000 µg/ml in methanol
- Pk:1
- CAS No.:94-82-6
- Description:2,4-DB
- Formula:C₁₀H₁₀Cl₂O₃
- Description:2,4-DB
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.