Pesticide reference standard, Simazine
Catalog # REST32236
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml in acetone
- Pk:1
- CAS No.:122-34-9
- Description:Simazine
- Formula:C₇H₁₂ClN₅
- Description:Simazine
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.