β-BHC
β-BHC, 1,000 µg/ml in acetone, 1 ml/ampoule, 1
Catalog # REST32209
Supplier: Restek
Jump to:
Product Details & Documents
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Concentration:1,000 µg/ml in acetone
- Pk:1
- Description:β-BHC
- Pk Info:1 ml/ampoule
- Cat. No.:REST32209
- Formula:C₆H₆Cl₆
- Description:β-BHC
- CAS No.:319-85-7
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



