ContactUsLinkComponent
Adapter, Dino-Lite FC-L-MA1
Adapter, Dino-Lite FC-L-MA1
  630-1817
undefined
Adapter, Dino-Lite FC-L-MA1
  630-1817
  • Description:
    Mirror angle adapter, Dino-Lite FC-L-MA1, 68×7 mm, 45°, for all AD models with long working distance
  • Seq:
    920
  • Pk:
    1
This mirror angle adapter fits onto the models with long working distance: AD4113TL/AD4116TL/AD4112PTL/AD4013MTL/AD4013MZTL/AD7013MTL and more.
Mirror angle: 45 °