Heavy duty multi purpose cleaner, TASKI Sprint Spitfire Plus
Catalog # DIVY7513489
Supplier: Diversey
Specifications
- Description:Heavy duty multi purpose cleaner, TASKI Sprint Spitfire Plus
- Pack type:Spray bottle
- Packed:6×750 ml
- Pk:6
Specifications
About this item
Ready to use, alkaline heavy-duty cleaner for most alkaline resistant, hard surfaces.
- Alkaline and solvent based formulation
- Good emulsifying power
- No rinse, streak-free
- Leaves a pleasant fragrance
- Unique, patented Odour Neutralizing Technology (O.N.T.)
Simultaneously removes grease and oils, particularly efficient for stain removal such as ink, marker, pencil, lipstick, black heel marks, smoke films, greasy food and other tenacious stains.