Orders LinkContactUsLinkComponent
Label printer, colour, J2000
Label printer, colour, J2000
Label printer, colour, J2000
Supplier:  
Certificates
Label printer, colour, J2000
Label printer, colour, J2000
0 Options Available Below
Product Details & Documents
The BradyJet J2000 is a practical and easy to use, full colour inkjet label printer that can print reliable, high resolution signs, facility and laboratory sample labels. Suitable for many applications including lean/5S applications and die-cut materials for wire, laboratory or general identification, GHS / CLP labels, pipe markers, equipment labels, inventory identification, tags, tube labels, cryo labels and glassware labels.
  • High, photo quality printing resolution of 4800 dpi
  • Quality images, barcodes and pictures can be added to any sign
  • Easy colour coding as can print in any colour
  • Print speed of 63 mm/second enables larger colour coded labels to be printed on demand quickly
Product Family Options
Labels for BradyJet J2000 Lab printer
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-59-7425J
    B-7425J
    PP
    Printer labels
    2000 labels/roll
    25×12 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 164,60 €
    164,60 €
    1 * 1 Roll
       164,60 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-257-7425J
    B-7425J
    PP
    Printer labels
    1500 labels/roll
    25×9 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 180,39 €
    180,39 €
    1 * 1 Roll
       180,39 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-137-7425J
    B-7425J
    PP
    Printer labels
    1500 labels/roll
    50×25 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 123,25 €
    123,25 €
    1 * 1 Roll
       123,25 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-249-7425J
    B-7425J
    PP
    Labels with 9 mm dot
    1500 labels/roll
    25×9 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 186,00 €
    186,00 €
    1 * 1 Roll
       186,00 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-53-7425J
    B-7425J
    PP
    Printer labels
    2000 labels/roll
    50×12 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 216,03 €
    216,03 €
    1 * 1 Roll
       216,03 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-179-7425J
    B-7425J
    PP
    Printer labels
    1500 labels/roll
    25×25 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 162,02 €
    162,02 €
    1 * 1 Roll
       162,02 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-258-7425J
    B-7425J
    PP
    Printer labels
    900 labels/roll
    38×44 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 155,70 €
    155,70 €
    1 * 1 Roll
       155,70 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-251-7425J
    B-7425J
    PP
    Labels with 11 mm dot
    1500 labels/roll
    25×12 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 178,47 €
    178,47 €
    1 * 1 Roll
       178,47 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-152-7425J
    B-7425J
    PP
    Printer labels
    2000 labels/roll
    25×9 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 151,19 €
    151,19 €
    1 * 1 Roll
       151,19 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-256-7425J
    B-7425J
    PP
    Printer labels
    1500 labels/roll
    33×15 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 147,81 €
    147,81 €
    1 * 1 Roll
       147,81 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-136-7425J
    B-7425J
    PP
    Printer labels
    1500 labels/roll
    38×19 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 228,61 €
    228,61 €
    1 * 1 Roll
       228,61 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-235-7425J
    B-7425J
    PP
    Printer labels
    1250 labels/roll
    38×25 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 97,80 €
    97,80 €
    1 * 1 Roll
       97,80 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-37-7425J
    B-7425J
    PP
    Printer labels
    1500 labels/roll
    38×12 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 157,54 €
    157,54 €
    1 * 1 Roll
       157,54 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-151-7425J
    B-7425J
    PP
    Printer labels
    2000 labels/roll
    50×6 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 149,44 €
    149,44 €
    1 * 1 Roll
       149,44 €
Product Information
  • Catalog #
    Printer model
    Model
    Type
    Material
    Description
    Packed
    Dimensions
    Colour
    Availability
    Price
  • J2000-Lab
    J20-88-7425J
    B-7425J
    PP
    Printer labels
    2000 labels/roll
    25×19 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 321,33 €
    321,33 €
    1 * 1 Roll
       321,33 €
Labels for BradyJet J2000 SFID printer (Inkjet receptive polyester)
Product Information
  • Catalog #
    Printer model
    Dimensions
    Description
    Material
    Type
    Model
    Packed
    Printing method
    Colour
    Availability
    Price
  • BradyJet J2000 SFID
    75×110 mm
    Printer labels
    Polyester
    B-2569
    J20-261-2569
    270 labels/roll
    Inkjet
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 347,74 €
    347,74 €
    1 * 1 Roll
       347,74 €
Product Information
  • Catalog #
    Printer model
    Dimensions
    Description
    Colour
    Material
    Type
    Model
    Packed
    Printing method
    Availability
    Price
  • 026
    Inkjet
    J20C-4000-2569
    BradyJet J2000 SFID
    Label printer tape
    101 mm×30 m
    B-2569
    Polyester
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 391,03 €
    391,03 €
    1 * 1 Roll
       391,03 €
Product Information
  • Catalog #
    Printer model
    Dimensions
    Description
    Colour
    Material
    Type
    Model
    Packed
    Printing method
    Availability
    Price
  • 024
    Inkjet
    J20C-1125-2569
    BradyJet J2000 SFID
    Label printer tape
    29 mm×30 m
    B-2569
    Polyester
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 204,57 €
    204,57 €
    1 * 1 Roll
       204,57 €
Product Information
  • Catalog #
    Printer model
    Dimensions
    Description
    Material
    Type
    Model
    Packed
    Printing method
    Colour
    Availability
    Price
  • BradyJet J2000 SFID
    55×85 mm
    Printer labels
    Polyester
    B-2569
    J20-260-2569
    340 labels/roll
    Inkjet
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 315,28 €
    315,28 €
    1 * 1 Roll
       315,28 €
Product Information
  • Catalog #
    Printer model
    Dimensions
    Description
    Material
    Type
    Model
    Packed
    Printing method
    Colour
    Availability
    Price
  • BradyJet J2000 SFID
    105×155 mm
    Printer labels
    Polyester
    B-2569
    J20-262-2569
    200 labels/roll
    Inkjet
    White
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 480,62 €
    480,62 €
    1 * 1 Roll
       480,62 €
Product Information
  • Catalog #
    Printer model
    Dimensions
    Description
    Colour
    Material
    Type
    Model
    Packed
    Printing method
    Availability
    Price
  • 025
    Inkjet
    J20C-2250-2569
    BradyJet J2000 SFID
    Label printer tape
    57 mm×30 m
    B-2569
    Polyester
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 306,18 €
    306,18 €
    1 * 1 Roll
       306,18 €
Labels for BradyJet J2000 SFID printer (Inkjet receptive vinyl)
Product Information
  • Catalog #
    Printing method
    Model
    Colour
    Printer model
    Description
    Dimensions
    Type
    Material
    Availability
    Price
  • Inkjet
    J20C-1125-2595
    White
    BradyJet J2000 SFID
    Label printer tape
    29 mm×30 m
    B-2595
    Vinyl
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 176,52 €
    176,52 €
    1 * 1 Roll
       176,52 €
Product Information
  • Catalog #
    Printing method
    Model
    Colour
    Printer model
    Description
    Dimensions
    Type
    Material
    Availability
    Price
  • Inkjet
    J20C-2250-2595
    White
    BradyJet J2000 SFID
    Label printer tape
    57 mm×30 m
    B-2595
    Vinyl
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 261,30 €
    261,30 €
    1 * 1 Roll
       261,30 €
Product Information
  • Catalog #
    Printing method
    Model
    Colour
    Printer model
    Description
    Dimensions
    Type
    Material
    Availability
    Price
  • 55×85 mm
    Printer labels
    028
    Vinyl
    B-2595
    J20-260-2595
    340 labels/roll
    Inkjet
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 285,98 €
    285,98 €
    1 * 1 Roll
       285,98 €
Product Information
  • Catalog #
    Printing method
    Model
    Colour
    Printer model
    Description
    Dimensions
    Type
    Material
    Availability
    Price
  • Inkjet
    J20C-4000-2595
    White
    BradyJet J2000 SFID
    Label printer tape
    101 mm×30 m
    B-2595
    Vinyl
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 361,00 €
    361,00 €
    1 * 1 Roll
       361,00 €
Product Information
  • Catalog #
    Printing method
    Model
    Colour
    Printer model
    Description
    Dimensions
    Type
    Material
    Availability
    Price
  • 75×110 mm
    Printer labels
    029
    Vinyl
    B-2595
    J20-261-2595
    270 labels/roll
    Inkjet
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 315,45 €
    315,45 €
    1 * 1 Roll
       315,45 €
Product Information
  • Catalog #
    Printing method
    Model
    Colour
    Printer model
    Description
    Dimensions
    Type
    Material
    Availability
    Price
  • 105×155 mm
    Printer labels
    030
    Vinyl
    B-2595
    J20-262-2595
    200 labels/roll
    Inkjet
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 435,98 €
    435,98 €
    1 * 1 Roll
       435,98 €
Labels for BradyJet J2000 SFID printer (Polyester with inkjet receptive vinyl topcoat)
Product Information
  • Catalog #
    Printing method
    Model
    Colour
    Printer model
    Description
    Dimensions
    Type
    Material
    Availability
    Price
  • Inkjet
    J20C-3250-2551
    White
    BradyJet J2000 SFID
    Label printer tape
    83 mm×15 m
    B-2551
    Polyester/vinyl
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 191,60 €
    191,60 €
    1 * 1 Roll
       191,60 €
Product Information
  • Catalog #
    Printing method
    Model
    Colour
    Printer model
    Description
    Dimensions
    Type
    Material
    Availability
    Price
  • Inkjet
    J20C-2500-2551
    White
    BradyJet J2000 SFID
    Label printer tape
    64 mm×15 m
    B-2551
    Polyester/vinyl
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 151,42 €
    151,42 €
    1 * 1 Roll
       151,42 €
Product Information
  • Catalog #
    Printing method
    Model
    Colour
    Printer model
    Description
    Dimensions
    Type
    Material
    Availability
    Price
  • 82×146 mm
    Printer labels
    040
    Polyester/vinyl
    B-2551
    J20-255-2551
    100 labels/roll
    Inkjet
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 317,33 €
    317,33 €
    1 * 1 Roll
       317,33 €