Herbicide mix #2, derivatised form
Supplier: Restek
Certificates
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Specifications
- CAS No.:17640-02-7
- Formula:CH₃CCl₂COOCH₃
- Description:Dalapon methyl ester