Appendix IX mix #2
Catalog # REST31806
Supplier: Restek
CAS Number:
Specifications
- Pk:1 Ampoul
- Description:Appendix IX Mix #2 (32 components),
1000 µg/ml each in methylene chloride - Environmentally Preferable:
- Pk Info:1 ml/ampoule
- Type:EPA Appendix IX
Specifications
About this item
Appendix IX mix, contains 32 components.