Orders LinkContactUsLinkComponent
Grid Storage Box
Grid Storage Box
Supplier:   Merck
Grid Storage Box
Supplier:  Merck
Certificates
Grid Storage Box
2 Options Available Below
Product Details & Documents
This storage box is made up of plastic.
  • Anti-static
  • Record card is used to record identity of samples
  • Device is anti-static to prevent handling problems due to repulsion or attraction
Product Family Options
Product Information
  • Availability
    Price
  • SIALG6276-1EA
    Grid storage box
    Grid storage box
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 48,30 €
    48,30 €
    1 * 1 items
       48,30 €
  • SIALG6276-10EA
    Grid storage box
    Grid storage box
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 400,00 €
    400,00 €
    1 * 10 items
       400,00 €