ContactUsLinkComponent
Sharps containers, Clinisafe
Sharps containers, Clinisafe
Catalog # HUARHOS-30LPBBYLW
Supplier:  Hospidex
Sharps containers, Clinisafe
Catalog # HUARHOS-30LPBBYLW
Supplier:  Hospidex
Specifications
  • Capacity:
    30 L
  • Colour:
    Yellow/white
  • Seq:
    01
  • W×D×H:
    300x326x400 mm
  • Pk:
    1
Specifications
About this item
PP, yellow base with white lid. To collect hazardous medical waste. Lid with integrated handle. Single flap opening
  • Hinges that are very easy to assemble
  • Hinges allowing the lid to be tipped back fully, so the opening is completely clear
  • Original lid suspension hook system for use without hinges
  • Double position, temporary and permanent closure
  • Permanent closure lock system using 14 tabs which click in any other
  • Four permanent closure visual indicators
  • Original watertight gasket made from expanded PU
Marked with moisture-resistant label, which cannot be removed (ADR UN 3291-compliant). Carrying handles on two sizes. Handles with finger protection system. Suitable for one-handed use.
Label text in french and dutch with instructions for use
Certifications: Compliant with ADR, NFX 30-511 and ISO 23907 standards.