Genomic DNA of E. coli O157 (EDL 933), IRMM® certified Reference Material
Catalog # SIALIRMM449
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Description:Genomic DNA of E. coli O157
Specifications
About this item
Certified for genomic DNA Eschericha coli O157, strain EDL 933 confirmed by fliC gene.