Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lights
UV Lights, Lamps and Bulbs
UV lamp for Thermo HPLC system
UV lamp for Thermo HPLC system
UV lamp for Thermo HPLC system
Catalog # REST25484
Supplier:
Restek
Expand image
UV lamp for Thermo HPLC system
Catalog # REST25484
Supplier:
Restek
Specifications
For:
Spectrachrom 200, UV100, UV1000, UV150, UV200, UV2000 and UV3000
Description:
UV lamp
Environmentally Preferable:
Pk:
1
Specifications
About this item
Product Details
Product Details