Orders LinkContactUsLinkComponent
Precision thermometers
Precision thermometers
Supplier:   Amarell
Precision thermometers
Supplier:  Amarell
Certificates
Precision thermometers
2 Options Available Below
Product Details & Documents
Melting point thermometers according to Kofler.
  • Round top finish in transparent square plastic case
  • Durable pigment
  • Solid stem
These thermometers are suitable for government verification.
Product Family Options
Product Information
  • Range
    Division
    Length
    Filling
    Ø
    Availability
    Price
  • AMARM250470-FL
    Precision thermometer, white
    Precision thermometer, white
    +20…+230 °C
    1,0 °C
    250 mm
    Red
    6,0 - 6,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • AMARM250472-GA
    Precision thermometer, white
    Precision thermometer, white
    +120…+350 °C
    1,0 °C
    250 mm
    Red
    6,0 - 6,5 mm