ContactUsLinkComponent
Disposable particle filtering respirators, P391
  PRTWP391WHR
 :  Portwest
undefined
Disposable particle filtering respirators, P391
  PRTWP391WHR
 :  Portwest
  • Type:
    P391
  • Pk:
    10
  • With valve:
    +
  • Class:
    FFP3
Eagle FFP3 folding white mask with seven dynamic parts provides excellent facial movement with maximum comfort. Valved and with inner foam to increase the comfort of the wearer.
  • High performance exhalation valve to reduce heat and provide comfort when used in hot and humid conditions
  • Large inner space to help the user to breathe more easily
  • Flexible shape which allows the mask to be folded flat
Dolomite optional clogging test (D) to provide more comfortable breathing and longer lasting performance of filtration.
 : EN 149 FFP3 (NR D)
 : Individually wrapped and packed inside hygienic polybag for vending machines.