Orders LinkContactUsLinkComponent
Objectives for CKX53
Supplier:   OLYMPUS OPTICAL
Objectives for CKX53
Supplier:  OLYMPUS OPTICAL
Certificates
Objectives for CKX53
Objectives for CKX53
4 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Magnification
    Working distance
    Availability
    Price
  • OLYMN5229300
    Dry objective with pre-centre phase contrast slider
    Dry objective with pre-centre phase contrast slider
    Ph 20×/0,4 (dry)
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 743,00 €
    743,00 €
    1 * 1 items
       743,00 €
  • OLYMN5229400
    Dry objective with pre-centre phase contrast slider
    Dry objective with pre-centre phase contrast slider
    Ph 40×/0,55
    2,2 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 724,00 €
    724,00 €
    1 * 1 items
       724,00 €
  • OLYMN4239000
    Plan achromatic objective
    Plan achromatic objective
    10×/0,25
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 197,00 €
    197,00 €
    1 * 1 items
       197,00 €
  • OLYMN4239600
    C plan achromatic objective
    C plan achromatic objective
    10×/0,17
    10,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 373,00 €
    373,00 €
    1 * 1 items
       373,00 €