ContactUsLinkComponent
Deep well V-bottom plates, 24-well
Deep well V-bottom plates, 24-well
Catalog # SARPA-0038
Supplier:  Sartorius
Deep well V-bottom plates, 24-well
Catalog # SARPA-0038
Supplier:  Sartorius
Specifications
  • Packed:
    Individually wrapped
  • Sterile:
    +
  • Colour:
    Amber
  • Description:
    Deep 24-well V-bottom plates
  • Pk:
    1
  • Well volume:
    8 ml
Specifications
About this item
These 24-well deep well plates can be used to hold samples harvested from bioreactors. They can also be used to hold reagents and feeds on the storage deck, before being added into the bioreactor.
  • Irradiated
Delivery: Pack contains 25 sterile plates with lids, individually wrapped.