SPE columns, drugs of abuse analysis, CHROMABOND® Drug
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® Drug is a special silica phase for the SPE enrichment of drugs of abuse from urine or plasma.
- Base material: Silica
- Special bifunctional modification: C8 / SA (strong cation exchanger)
Application: Enrichment of acidic, neutral and basic drugs from urine or plasma.
Specifications
- Catalog No:
- MANA730168.250
- MANA738161.100M
- MANA730168
- MANA732168
- MANA730681
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g