SPE columns, CHROMABOND® HLB
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® HLB is a hydrophilic-lipophilic balanced N-vinylpyrrolidone-divinylbenzene resin for SPE.
- Applicable for a wide range of analyte polarities
- Enhanced retention for polar compounds
- High loadability and outstanding performance
- Water wettable – even if bed runs dry, SPE can be continued
Applications: Polar organic molecules from polar matrices e.g., sulphonamides, pesticides, chloramphenicol, iodinated contrast media.
Specifications
- Catalog No:
- 705-1108
- MANA730927
- MANA730922
- MANA732140
- MANA730922P30
- MANA732141
- MANA731923
- 705-1107
- MANA730923P30
- MANA730925
- MANA731921
- MANA730924.250
- MANA730924P30
- MANA730921P30
- MANA730921
- MANA730923.250
- MANA730923
- MANA730944
- MANA730944P30
- MANA738921.010M
- MANA738920.060M
- MANA738921.030M
- MANA731922
- MANA730929
- MANA730926.250
- MANA730928
- MANA730927.250
- pH range:
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- Surface area:
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g
- 750 m²/g