Orders LinkContactUsLinkComponent
Electrolyte for reference electrodes
LiCl solution in 1 mol/l in ethanol, 6×30 ml, 6
Catalog # METT51350088
Supplier:   Mettler - Toledo
Electrolyte for reference electrodes
Catalog # METT51350088
Supplier:  Mettler - Toledo
CAS Number:  
Electrolyte for reference electrodes
LiCl solution in 1 mol/l in ethanol, 6×30 ml, 6
CAS Number:  
Price
164,00 €
Log in for availability
1 * 6 items
Product Details & Documents
Used for the measurement in non-aqueous media.
Specifications
Specifications
  • Pk:
    6
  • Description:
    LiCl solution in 1 mol/l in ethanol, 6×30 ml
Specifications