Orders LinkContactUsLinkComponent
Multi-element quality control standards for ICP
Multi-element quality control standards for ICP
Supplier:   CPACHEM
Multi-element quality control standards for ICP
Supplier:  CPACHEM
Certificates
Multi-element quality control standards for ICP
Multi-element quality control standards for ICP
2 Options Available Below
Product Details & Documents
ICP QC standards used for Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr, Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Ni, Nb, Pb, Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn. 1 mg/l for 33 elememts.
Documents
Product Family Options
Product Information
  • Matrix
    Pk
    Availability
    Price
  • ACSDM8A96.1.5N.L1
    Multi-element quality control standards for 33 elements
    Multi-element quality control standards for 33 elements
    5% HNO₃
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 148,00 €
    148,00 €
    1 * 100 mL
       148,00 €
  • ACSDM8A96.1.5N.L5
    Multi-element quality control standards for 33 elements
    Multi-element quality control standards for 33 elements
    5% HNO₃
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 338,00 €
    338,00 €
    1 * 500 mL
       338,00 €