ContactUsLinkComponent
Surgical tapes, Durapore™
  MMMEM1538-1
Surgical tapes, Durapore™
  MMMEM1538-1
  • Description:
    Surgical tape, white
  • L×W:
    9,1 m × 25,0 mm
  • Seq:
    02
  • Pk:
    12 Roll
Durable, no-stretch silk-like cloth tape for critical tubing applications. Silk-like cloth, hypoallergenic tape, standard roll.
  • High strength, versatile, conformable
  • Adheres well to dry skin
  • Convenient bidirectional tear
  • Hypoallergenic and not made with natural rubber latex
  • Water resistant
Adhesion coupled with a strong backing with high load bearing capacity makes it ideal for securing critical tubings and devices and stabilising finger splints.