Orders LinkContactUsLinkComponent
Labels, pressure-sensitive
Labels, pressure-sensitive
Labels, pressure-sensitive
Labels, pressure-sensitive
Labels, pressure-sensitive
These Nalgene™ polyolefin pressure-sensitive labels are used with photocopiers, laser printers or inkjet printers. Accepts a variety of inks including copier, marker, ballpoint, gels, and technical markers (pretest specific inks for suitability prior to use), does not accept pencil.
  • Rated to −29 °C
  • Resistant to ultraviolet light, mildew, chemicals and dirt and won't discolor, crack or tear
  • Excellent for labeling microscopic slides - adhesive backing sticks well to any plastic, glass or metal surface
  • Chemically compatible with most lab reagents although solvents may dissolve adhesive
  • Autoclavable
Formatted for easy setup in word processing programs; instructions included in each pack.
  • Catalog No:
  • 817-0081
  • 817-0082
  • 817-0083
  • 817-0084
  • 817-0085
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Product Family Options
  • Description
    Material
    Colour
    Dimensions
  • Labels
    Polyolefin
    White
    22×22 mm
    1 * 800 items
    1 * 800 items - 84,90 €
    84,90 €
    1 * 800 items
       84,90 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    817-0081
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Labels
    Polyolefin
    White
    20×38 mm
    1 * 650 items
    1 * 650 items - 91,40 €
    91,40 €
    1 * 650 items
       91,40 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    817-0082
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Labels
    Polyolefin
    White
    25×67 mm
    1 * 300 items
    1 * 300 items - 76,20 €
    76,20 €
    1 * 300 items
       76,20 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    817-0083
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Labels
    Polyolefin
    White
    13×44 mm
    1 * 800 items
    1 * 800 items - 97,30 €
    97,30 €
    1 * 800 items
       97,30 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    817-0084
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Labels
    Polyolefin
    White
    51×102 mm
    1 * 100 items
    1 * 100 items - 84,90 €
    84,90 €
    1 * 100 items
       84,90 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    817-0085
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact