ContactUsLinkComponent
12-Crown-4 (1,4,7,10-tetraoxacyclododecane, lithium ionophore V)
12-Crown-4 (1,4,7,10-tetraoxacyclododecane, lithium ionophore V)
Catalog # MOLE16255417-5G
Supplier:  Molekula
CAS Number:  
12-Crown-4 (1,4,7,10-tetraoxacyclododecane, lithium ionophore V)
Catalog # MOLE16255417-5G
Supplier:  Molekula
CAS Number:  
Specifications
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
Specifications
About this item
 :
 :176.21 g/mol
Boiling Pt:254 °C (1013 hPa)
Melting Pt:16 °C
Density:1.11 g/cm³ (20 °C)
EINECS:206-036-5
UN:2810
ADR:6.1,II
CAS Number:294-93-9
Formula:C₈H₁₆O₄
MDL Number:MFCD00005103