VWR®, Accessories for VWR® Semi-micro, Analytical or Precision Balances
611-3864
- Description:Ioniser panel, allows user to retrofit an integrated ioniser
- Seq:311
- Pk:1
- For:VWR semi-micro, analytical or precision balances fitted with draftshields which have 3 sliding doors