Cesium perchlorate, Reagent Grade
Catalog # 12308.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
:
:232.36 g/mol
EINECS:236-643-0
UN:1481
:GHS03
:GHS07
Storage Temperature:Ambient
CAS Number:13454-84-7
Formula:ClCsO₄
MDL Number:MFCD00010956
Melting Pt:250 °C (dec.)
Density:3.32 g/cm³ (20 °C)
ADR:5.1 II