ContactUsLinkComponent
4-Chlorobiphenyl-[13C12] (PCB No. 3) 40 µg/ml in n-nonane (99% 13C), CIL
4-Chlorobiphenyl-[13C12] (PCB No. 3) 40 µg/ml in n-nonane (99% 13C), CIL
Catalog # PROOCILEC-4990-1.2
CAS Number:  
4-Chlorobiphenyl-[13C12] (PCB No. 3) 40 µg/ml in n-nonane (99% 13C), CIL
Catalog # PROOCILEC-4990-1.2
CAS Number:  
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • Description:
    4-Chlorobiphenyl-[13C12] (PCB No. 3) 40 µg/ml in n-nonane (99% 13C)