2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156)
Catalog # PROOCIL-PCB-156-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:360.88 g/mol
Storage Temperature:Refrigerator
UN:3432
ADR:9,II
CAS Number:38380-08-4
Formula:C₁₂H₄Cl₆