1,4-Naphthoquinone
Catalog # MOLE25038144-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:158.16 g/mol
Merck Index:12,06482
Boiling Pt:298 °C (1013 hPa)
Melting Pt:124 °C
Density:1.422 g/cm³ (20 °C)
Flash Pt:141 °C
EINECS:204-977-6
UN:2811
ADR:6.1,I
:GHS06
:GHS09
CAS Number:130-15-4
Formula:C₁₀H₆O₂
MDL Number:MFCD00001676