J.T.Baker® BAKERBOND Speedisk® Amino (NH2), SPE Columns
8165-03
: Avantor
- Capacity:3 ml
- Bed weight:35 mg
- Phase:Amino (NH₂)
- Pk:1
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate prefiltration and concentration steps.
- Run times up to nine times faster than conventional SPE columns
- Unique laminar configuration
- Silica based
General applications: Lipids (fatty acids, cholesterol).
These SPE columns can be used in ion exchange and normal phase mode.