1-Hexanesulphonic acid sodium salt, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2832-45-3
:MFCD00007542
Formula:C₆H₁₃O₃SNa
Melting Pt:300 °C
:M-Clarity™ quality level = MQ300
:188.22 g/mol
Storage Temperature:Ambient
EINECS:220-601-3
Frequently Bought Together