Orders LinkContactUsLinkComponent
N,N-Dimethylformamide
Flammable
Health
Irritant
Supplier:   Roth Carl
N,N-Dimethylformamide
Supplier:  Roth Carl
Certificates
N,N-Dimethylformamide
N,N-Dimethylformamide
3 Options Available Below
Product Details & Documents
Synonyms:  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  • CAS Number:68-12-2
  • Formula:HCON(CH₃)₂
  • MDL Number:MFCD00003284
  • Molecular Weight:73.09 g/mol
  • Merck Index:13,03269
  • Boiling Pt:153 °C (1013 hPa)
  • Melting Pt:–61 °C
  • Density:0.949 g/cm³ (20 °C)
  • Flash Pt:57.5 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-679-5
  • UN:2265
  • ADR:3,III
  • REACH:01-2119475605-32
Product Family Options
Product Information
  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 51,80 €
    51,80 €
    1 * 1 L
       51,80 €
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 104,00 €
    104,00 €
    1 * 2,5 L
       104,00 €
  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 591,00 €
    591,00 €
    1 * 25 L
       591,00 €