Organic reference standard, Chloroacetonitrile 1000 µg/ml in methanol
Catalog # PROOU-EPA-1205
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:1
- CAS No.:107-14-2
- Description:Chloroacetonitrile 1,000 µg/ml in methanol
- Formula:C₂H₂ClN
- Description:Chloroacetonitrile