Organic reference standard, 2,3'-Dichlorobiphenyl (PCB No. 6) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-6
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:25569-80-6
- Description:2,3'-Dichlorobiphenyl (PCB No. 6) 35 µg/ml in isooctane
- Formula:C₁₂H₈Cl₂
- Description:2,3'-Dichlorobiphenyl (PCB No. 6)