Organic reference standard, 1,1,1-Trichloroethane 100 µg/ml in methanol
Catalog # PROOU-HC-250-1
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:1
- CAS No.:71-55-6
- Description:1,1,1-Trichloroethane 100 µg/ml in methanol
- Formula:C₂H₃Cl₃
- Description:1,1,1-Trichloroethane