Detergents for glasswares, NOCHROMIX®, Sigma-Aldrich®
SIAL328693-10PAK
: Merck
- Description:NOCHROMIX® cleaning agent
- Environmentally Preferable:
- Pk:10
A metal-free white powder when mixed with sulfuric acid, makes 2,3 l of cleaning solution for laboratory glasswares.
- Non-polluting
- Non-carcinogenic
: EPA recommended.