Orders LinkContactUsLinkComponent
Agar
Agar
Glass bottle for solids, 250 g
Catalog # ROTH5210.1
Supplier:   Roth Carl
Agar
Catalog # ROTH5210.1
Supplier:  Roth Carl
CAS Number:  9002-18-0
Agar
Agar
Glass bottle for solids, 250 g
CAS Number:  9002-18-0
Price
49,80 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications
Specifications
  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g