ContactUsLinkComponent
Abdominal spatula, type Haberer, Aesculap®
Abdominal spatula, type Haberer, Aesculap®
Catalog # AESCBT758R
Supplier:  Aesculap
Abdominal spatula, type Haberer, Aesculap®
Catalog # AESCBT758R
Supplier:  Aesculap
Specifications
  • Length:
    305 mm
  • Seq:
    05
  • Blade width:
    50/40 mm
  • Pk:
    1
Specifications
About this item
This non sterile, reusable stomach spatula has a tapered, malleable form.