Order Entry
Slovenia
ContactUsLinkComponent
 
Nickel standard solution, 10000 mg/L Ni in dil. nitric acid standard for AAS
Catalog # PROOU-IAA-028
CAS Number:  
Nickel standard solution, 10000 mg/L Ni in dil. nitric acid standard for AAS
Catalog # PROOU-IAA-028
CAS Number:  

Specifications

  • Concentration:
    10000mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Element:
    Nickel
  • Symbol:
    Ni

Specifications

About this item